Learn More
MORE DIAGNOSTICS INC Level-1 Rap/Tac/CsA Control | 6x4ml
SDP
Catalog No. NC1751485
Encompass
A stable assayed control for the monitoring of accuracy and precision of Rapamycin (Sirolimus), tacrolimus and Cyclosporine A for immunoassays and LC-MS/MS quantitative methods.
Catalog No. NC1751485 Supplier MORE DIAGNOSTICS INC Supplier No. 2901
A stable assayed control for the monitoring of accuracy and precision of Rapamycin (Sirolimus), tacrolimus and Cyclosporine A for immunoassays and LC-MS/MS quantitative methods.