Learn More
Premier Lab Supply CEMBLES Pre-Assembled XRF Sample Cups – PN# PA-460-45-240
SDP
Catalog No. NC1824610
Encompass_Preferred
CEMBLES Pre-Assembled XRF Sample Cups! (CEMBLES PN# PA-460-45-240; 45mm Cups PN# SC-3345; Polypropylene film 4.0μm; 32 per pack). CEMBLES are custom made to your specifications & available in variant configurations of cups and thin films that have been pre-assembled for your convenience.
This item is not returnable. View return policy
Catalog No. NC1824610 Supplier Premier Lab Supply Supplier No. PA46045240
CEMBLES Pre-Assembled XRF Sample Cups! (CEMBLES PN# PA-460-45-240; 45mm Cups PN# SC-3345; Polypropylene film 4.0μm; 32 per pack). CEMBLES are custom made to your specifications & available in variant configurations of cups and thin films that have been pre-assembled for your convenience.