Learn More
Waters Corp Waters Total Recovery Vial, Clear Glass, 12 x 32 mm, Screw Neck
Catalog No. 50791169 Shop All Waters Corporation Products
Encompass_Preferred
The clear glass 12 x 32 mm screw neck total recovery vial comes preassembled with a PTFE/silicone septum and is available in a 100-pack. It is designed as an autosampler vial with a 1 mL volume capacity. This product is suitable for both liquid and solid sample storage, making it versatile for various laboratory applications.
  • Preassembled with a cap and septa for convenience and a proper seal.
  • Designed for total sample recovery.
  • Suitable for storing both liquid and solid samples.
  • Screw neck design is compatible with standard laboratory equipment for easy and secure sealing.
  • Made of clear glass, resistant to a wide range of temperatures and most common laboratory chemicals.
  • 1 mL volume capacity.
  • 100 vials per pack.
Catalog No. 50-791-169 Supplier Waters Corp Supplier No. 186004631
The clear glass 12 x 32 mm screw neck total recovery vial comes preassembled with a PTFE/silicone septum and is available in a 100-pack. It is designed as an autosampler vial with a 1 mL volume capacity. This product is suitable for both liquid and solid sample storage, making it versatile for various laboratory applications.
  • Preassembled with a cap and septa for convenience and a proper seal.
  • Designed for total sample recovery.
  • Suitable for storing both liquid and solid samples.
  • Screw neck design is compatible with standard laboratory equipment for easy and secure sealing.
  • Made of clear glass, resistant to a wide range of temperatures and most common laboratory chemicals.
  • 1 mL volume capacity.
  • 100 vials per pack.