Learn More
Waters Corp WATERS CORPORATION Cartridge Assembly, Check Valve, 2 pk, for UPLC Systems
Catalog No. 502039474 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This cartridge assembly is designed as a crucial component of the Intelligent Intake Valve. It is specifically used with Binary Solvent Managers and Quaternary Solvent Managers, ensuring optimal performance for UPLC systems.
  • Functions as a check valve
  • Supplied in a package of 2 units
  • Compatible with specific Binary Solvent Managers and Quaternary Solvent Managers
  • Suitable for various UPLC system configurations, including H-Class, I-Class, and 2D systems
  • Replacement recommended as needed to maintain system integrity
Catalog No. 50-203-9474 Supplier Waters Corp Supplier No. 700003563
This cartridge assembly is designed as a crucial component of the Intelligent Intake Valve. It is specifically used with Binary Solvent Managers and Quaternary Solvent Managers, ensuring optimal performance for UPLC systems.
  • Functions as a check valve
  • Supplied in a package of 2 units
  • Compatible with specific Binary Solvent Managers and Quaternary Solvent Managers
  • Suitable for various UPLC system configurations, including H-Class, I-Class, and 2D systems
  • Replacement recommended as needed to maintain system integrity