Learn More
Waters Corp Waters Corporation Assembly, Extension Loop, 100μL, Bio, UPLC
Catalog No. 502039868 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is a replacement Assembly, Extension Loop, with a 100μL volume, specifically designed as an optional extension loop for an ACQUITY Arc Bio Sample Manager - FTN. It is categorized as a mechanical spare part for UPLC systems, to be replaced as required. The “Bio” designation indicates its intended use with biochemical or biological applications, specifically with the ACQUITY Arc Bio Sample Manager.
  • Replacement part
  • Volume: 100μL
  • Designed for ACQUITY Arc Bio Sample Manager - FTN
  • Type: Mechanical
  • System type: UPLC
  • Replacement schedule: As required
  • Intended for biochemical or biological applications
Catalog No. 50-203-9868 Supplier Waters Corp Supplier No. 700011705
This is a replacement Assembly, Extension Loop, with a 100μL volume, specifically designed as an optional extension loop for an ACQUITY Arc Bio Sample Manager - FTN. It is categorized as a mechanical spare part for UPLC systems, to be replaced as required. The “Bio” designation indicates its intended use with biochemical or biological applications, specifically with the ACQUITY Arc Bio Sample Manager.
  • Replacement part
  • Volume: 100μL
  • Designed for ACQUITY Arc Bio Sample Manager - FTN
  • Type: Mechanical
  • System type: UPLC
  • Replacement schedule: As required
  • Intended for biochemical or biological applications