Learn More
Waters Corp Waters Corporation Extension Loop Assembly, 250 μL, Stainless Steel
Catalog No. 502039334 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This extension loop is a 250μL sample loop designed for various sample manager modules.
  • 250μL sample loop
  • Constructed from stainless steel
  • Compatible with HPLC and UPLC system types
  • Serves as a sample loop or tubing
  • Packaged as one unit
  • Replacement as required
Catalog No. 50-203-9334 Supplier Waters Corp Supplier No. 430002007
This extension loop is a 250μL sample loop designed for various sample manager modules.
  • 250μL sample loop
  • Constructed from stainless steel
  • Compatible with HPLC and UPLC system types
  • Serves as a sample loop or tubing
  • Packaged as one unit
  • Replacement as required