Learn More
Waters Corp Needle Assembly, 30 μL, MP35N, PPS Guide
Catalog No. 502391750 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 30μL MP35N needle assembly with a PPS guide is a replacement component for specific analytical systems, ensuring reliable performance in various laboratory applications.
  • It is designed for use with an ACQUITY APC Sample Manager -FTN.
  • The assembly is compatible with ACQUITY Arc HPLC and ACQUITY Arc systems.
  • It supports both HPLC and UPLC system types.
  • Features a 30μL capacity.
  • Constructed from MP35N material.
  • Includes a PPS guide.
Catalog No. 50-239-1750 Supplier Waters Corp Supplier No. 700009578
This 30μL MP35N needle assembly with a PPS guide is a replacement component for specific analytical systems, ensuring reliable performance in various laboratory applications.
  • It is designed for use with an ACQUITY APC Sample Manager -FTN.
  • The assembly is compatible with ACQUITY Arc HPLC and ACQUITY Arc systems.
  • It supports both HPLC and UPLC system types.
  • Features a 30μL capacity.
  • Constructed from MP35N material.
  • Includes a PPS guide.