Learn More
Waters Corp Waters Corporation Pump Head Assembly, Stainless Steel, with Check Valves
Catalog No. 50793469 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Replacement Pump Head Assembly is an integral part of the 515, 1515, and 1525 series pumps. It is manufactured from high-quality stainless steel and includes the inlet/outlet check valve and the Clear-100 Plunger Seal already installed. The Pump Head undergoes rigorous testing to ensure superior performance.
  • Integral part of 515, 1515, and 1525 series pumps.
  • Manufactured from high-quality stainless steel.
  • Includes inlet/outlet check valve.
  • Includes Clear-100 Plunger Seal installed.
  • Rigorously tested for superior performance.
Catalog No. 50-793-469 Supplier Waters Corp Supplier No. WAS207006
The Replacement Pump Head Assembly is an integral part of the 515, 1515, and 1525 series pumps. It is manufactured from high-quality stainless steel and includes the inlet/outlet check valve and the Clear-100 Plunger Seal already installed. The Pump Head undergoes rigorous testing to ensure superior performance.
  • Integral part of 515, 1515, and 1525 series pumps.
  • Manufactured from high-quality stainless steel.
  • Includes inlet/outlet check valve.
  • Includes Clear-100 Plunger Seal installed.
  • Rigorously tested for superior performance.