Learn More
Beckman Coulter Beckman Coulter VFC 8.50 Fixed-Angle Rotor Assembly for Allegra V-15R (MPN C63139)
Catalog No. NC3716315
Encompass
The Beckman Coulter VFC 8.50 (MPN C63139) is a fixed-angle biosafe rotor designed for use with Allegra V-series benchtop centrifuges. It supports processing of conical tubes for pelleting and separation workflows in research and clinical labs.
  • Fixed-angle rotor design for conical tube applications.
  • Holds up to eight tubes for a 400 mL nominal capacity.
  • Accepts 15 mL, 20-30 mL, or 50 mL tubes with appropriate adapters.
  • Maximum speed 11,360 rpm (15,032 x g).
  • rMAX 104 mm, rMIN 43 mm, rAV 73.5 mm.
  • k-factor 1,731 for sedimentation efficiency reference.
  • Anodized aluminum construction for durability and corrosion resistance.
  • Biosafe containment design for safer sample handling.
Catalog No. NC3716315 Supplier Beckman Coulter Supplier No. C63139
The Beckman Coulter VFC 8.50 (MPN C63139) is a fixed-angle biosafe rotor designed for use with Allegra V-series benchtop centrifuges. It supports processing of conical tubes for pelleting and separation workflows in research and clinical labs.
  • Fixed-angle rotor design for conical tube applications.
  • Holds up to eight tubes for a 400 mL nominal capacity.
  • Accepts 15 mL, 20-30 mL, or 50 mL tubes with appropriate adapters.
  • Maximum speed 11,360 rpm (15,032 x g).
  • rMAX 104 mm, rMIN 43 mm, rAV 73.5 mm.
  • k-factor 1,731 for sedimentation efficiency reference.
  • Anodized aluminum construction for durability and corrosion resistance.
  • Biosafe containment design for safer sample handling.