Learn More
Beckman Coulter VS 2.5-96 swinging-bucket rotor for Allegra V-15R (Beckman Coulter C63145)
Catalog No. NC2602772
Encompass
The VS 2.5-96 swinging-bucket rotor (MPN C63145) is designed for the Allegra V-15R benchtop centrifuge to process microtiter and deepwell plates. It accepts multiple plate formats and is BioSafe-compatible when used with the required BioSafe covers.
  • Holds 2 x 5 microtiter plates or 1 x 2 deepwell plates.
  • Total rotor capacity 960 mL.
  • Maximum speed 5,400 rpm (120V) / 5,700 rpm (240V).
  • Maximum force 4,932 x g (120V) / 5,495 x g (240V).
  • BioSafe-compatible; BioSafe covers required for biocertification.
Catalog No. NC2602772 Supplier Beckman Coulter Supplier No. C63145
Add to Cart
Add to Cart
Request a Quote
The VS 2.5-96 swinging-bucket rotor (MPN C63145) is designed for the Allegra V-15R benchtop centrifuge to process microtiter and deepwell plates. It accepts multiple plate formats and is BioSafe-compatible when used with the required BioSafe covers.
  • Holds 2 x 5 microtiter plates or 1 x 2 deepwell plates.
  • Total rotor capacity 960 mL.
  • Maximum speed 5,400 rpm (120V) / 5,700 rpm (240V).
  • Maximum force 4,932 x g (120V) / 5,495 x g (240V).
  • BioSafe-compatible; BioSafe covers required for biocertification.