Learn More
Hach Company Replacement: SPE Filtration Assembly for Xenosep SPE Starter Kit
Catalog No. 501996923 Shop All Hach Company Products
Encompass
SPE Filtration Assembly includes: 1 x Funnel, 300 mL, 47 mm (2943217), 1 x Holder, Starter, 47 mm (2943216), 1 x Coupler, PTFE, 47 mm (2943218), 1 x Support, Stainless Steel, 47 mm (2943219), 1 x Clamp, Aluminum, 47 mm (2943220), 2 x O-Rings, FEP-Viton, Coupler, 47 mm (2943223).
Catalog No. 50-199-6923 Supplier Hach Company Supplier No. 2946704
SPE Filtration Assembly includes: 1 x Funnel, 300 mL, 47 mm (2943217), 1 x Holder, Starter, 47 mm (2943216), 1 x Coupler, PTFE, 47 mm (2943218), 1 x Support, Stainless Steel, 47 mm (2943219), 1 x Clamp, Aluminum, 47 mm (2943220), 2 x O-Rings, FEP-Viton, Coupler, 47 mm (2943223).