Learn More
Waters Corp Tubing Assembly, MP35N, 0.005 in ID, 10.5 in
Catalog No. 502392795 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Replacement Tube Assembly for PDA/TUV Detectors. This Tube Assembly is 10.5 in. long and has an internal diameter of 0.005 in.. It is made from MP35N and used in Bio ACQUITY systems.
  • Compatible with Bio ACQUITY systems
  • Made from MP35N
  • 10.5 in. length
  • 0.005 in. internal diameter
Catalog No. 50-239-2795 Supplier Waters Corp Supplier No. 700009097
Replacement Tube Assembly for PDA/TUV Detectors. This Tube Assembly is 10.5 in. long and has an internal diameter of 0.005 in.. It is made from MP35N and used in Bio ACQUITY systems.
  • Compatible with Bio ACQUITY systems
  • Made from MP35N
  • 10.5 in. length
  • 0.005 in. internal diameter