Learn More
Waters Corp Assembly, Tubing, SS with Gold Plated Ends, 0.007 in. ID, 22.5 in.
Catalog No. 502391345 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a stainless steel tubing assembly designed with gold-plated ends. It features an inner diameter of 0.007 in. and a total length of 22.5 in., serving as a replacement component in various applications.
  • Tubing is constructed from stainless steel
  • Features gold-plated ends for enhanced connectivity
  • Inner diameter measures 0.007 in.
  • Overall length is 22.5 in.
  • Functions as a spare part kit
Catalog No. 50-239-1345 Supplier Waters Corp Supplier No. 700009703
This product is a stainless steel tubing assembly designed with gold-plated ends. It features an inner diameter of 0.007 in. and a total length of 22.5 in., serving as a replacement component in various applications.
  • Tubing is constructed from stainless steel
  • Features gold-plated ends for enhanced connectivity
  • Inner diameter measures 0.007 in.
  • Overall length is 22.5 in.
  • Functions as a spare part kit