Learn More
DIAGNOSTICA STAGO INC Asserachrom IX:Ag ELISA Kit, Diagnosis of Factor IX Deficiency, 96 Tests/Kits
Supplier:  DIAGNOSTICA STAGO INC 00943
Encompass
The Diagnostica Stago Asserachrom IX:Ag is a quantitative ELISA assay tailored for the accurate assessment of Factor IX antigen in clinical specimens, crucial for diagnosing Hemophilia B. It includes all essential reagents, providing reliable results for clinical applications. With storage requirements of 2-8 °C and the ability to conduct 96 tests per kit, it ensures convenience and efficiency for healthcare professionals.
Catalog No. NC2157039