Description
- Featured Industry: Environmental Industrial Hygiene
Specifications
Specifications
| For Use With (Equipment) | LC-MS or LC-MS/MS |
| For Use With (Application) | Sampling Isocyanates |
| Quantity | 10/Pk. |
| Type | DBA-Impregnated Glass Fiber Filter |
Asset is a registered trademark of Sigma-Aldrich Co. LLC