Learn More
MilliporeSigma™ Supelco™ ASSET&Trade; EZ4-NCO Dry Sampler
Catalog No. 111010363 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
11-101-0363 1 Kit
1 options
Catalog No. 11-101-0363 Supplier MilliporeSigma™ Supelco™ Supplier No. 5047U
Add to Cart
Add to Cart
ASSET EZ4-NCO dry sampler kit contains 2 ASSET EZ4-NCO cartridges, 2 tubing adapters, and 1 calibration fitting. The kit makes it easy to get started sampling with ASSET - 1 cartridge for sampling and the other as a field blank.
TRUSTED_SUSTAINABILITY
Specifications
Description Kit (each kit contains: 2 sampling cartridges, 2 tubing adapters, 1 calibration fitting and datasheet)
Quantity 1 Kit
Asset is a registered trademark of Sigma-Aldrich Co. LLC