Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Associated Bag Biohazard Bag
SDP

Catalog No. 19804301 Shop All Associated Bag Company Products
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-804-301 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-804-301 Supplier Associated Bag Supplier No. 364100R
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Color Red
Includes Biohazard label
For Use With (Application) Safe for landfill or incineration
Thickness 2 mil
Product Type Biohazard Message Low Density Gusseted Liner
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.