Learn More
3M™ Assortment Pack Cable Tie CT06220
Catalog No. 19071024 Shop All 3M Products
Change view
Click to view available options
Material:
Nylon 6/6
Catalog No. Material
19-071-024 Nylon 6/6
1 options
Catalog No. 19-071-024 Supplier 3M™ Supplier No. 7000133471
  • $612.00
    $424.65 / Case of 1000
    Save $187.35
    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
3M™ Cable Ties are made with high quality 6/6 nylon and designed to secure wire bundles and harness components quickly without slipping.
  • Plenum Rated
  • Self Locking Head
  • Curved tip allows for faster threading and install
  • Pack contains 15 each of 3M standard natural nylon cable ties in 4-, 6-, 8- and 11-inch lengths plus 10 each of black, weather-resistant ties in 4-, 6-, 8-, and 11-inch lengths.
  • 3M Catalog# CT06220
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cable Tie
Model CT06220
Quantity 1000/Cs.
Color Assorted
Material Nylon 6/6
Length (English) Assorted