Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Assurance™ Grade Tellurium, 1,000 μg/mL, SPEX CertiPrep™
SDP

Catalog No. PLTE42X Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
30 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PLTE42X 500 mL
PLTE42M 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PLTE42X Supplier SPEX CertiPrep Supplier No. PLTE42X
Add to Cart
Add to Cart

Assurance™ Grade Tellurium is a single-element standard for AA and ICP analysis.

  • Made with acid and ASTM Type I Water.
  • Inorganic compounds and metals at 99.99% to 99.9999% purity.
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016.
  • Assurance™ grade certified reference materials (CRMs) are designed for atomic absorption (AA) and inductively coupled plasma (ICP) analysis.
  • Custom standards can be manufactured upon request.
  • Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Chemical Identifiers

CAS 13494-80-9 , 7732-18-5 , 7697-37-2
Molecular Formula Te
Molecular Weight (g/mol) 127.60
MDL Number MFCD00134062
InChI Key PORWMNRCUJJQNO-UHFFFAOYSA-N
IUPAC Name tellurium
SMILES [Te]

Specifications

Description Assurance™ Grade Tellurium, 1,000 μg/mL for AA and ICP in 10% HNO3; 500 mL
Packaging Bottle
Concentration 1000 μg/mL (1000 ppm)
Technique ICP, ICP-MS, AA, Ion Chromatography, XRF, Cation Measurement
UN Number UN2031
pH Not applicable
Melting Point Undetermined
Boiling Point 83°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Completely miscible
Solution Type Standard Solution
Type Standard
Solvent or Matrix 10% HNO3
Quantity 500 mL
Grade Assurance™ Grade
Chemical Name or Material Tellurium (Te)
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Corrosion
ShelfLife 12 months
DOTInformation Hazardous, Nitric acid solution, Class 8, Packing Group II
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.