Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARKRAY USA Assure™ Dose Control Solution

Catalog No. 22355166
Change view
Click to view available options
Quantity:
2 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-355-166 2 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-355-166 Supplier ARKRAY USA Supplier No. ARK 500006
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Assure™ Dose Control Solution with One bottle of Normal and one bottle of High control solution.

  • The Assure Dose Control Solution Normal has a blue cap, High has a green cap
  • Includes Normal and High bottles
  • For use with The Assure Platinum and Glucocard Vital Meters Only
  • For in vitro Diagnostic Use Only
  • Not intended for human consumption. Do not drink
  • To avoid contamination, NEVER touch tip of control solution bottle to test strip!
  • Dye in control solution may stain clothing or surfaces

Specifications

Certifications/Compliance FDA Blood Glucose Meter Standards
Concentration Normal/High
Format Tube
For Use With (Equipment) Assure Platinum
Type Contol Solution
Level 1, 2
Quantity 2 Pk.
Stability 36°F to 86°F
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.