Learn More
ARKRAY USA Assure™ Platinum Blood Glucose Meter
Catalog No. 22355169
Change view
Click to view available options
Type:
Blood Glucose Meter
Catalog No. Type
22-355-169 Blood Glucose Meter
1 options
This item is not returnable. View return policy
Catalog No. 22-355-169 Supplier ARKRAY USA Supplier No. ARK 500001
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
The Assure™ Platinum Blood Glucose Meter is FDA approved for multi-patient use. Designed with nurses and residents in mind. Small, 0.5 μL sample size, auto coding, LCD backlit display with large readout, and test strip ejector for touch free strip disposal.
Provides healthcare professionals with highly accurate test results. A small, 0.5 μL, sample is all that is needed for each test.
  • LCD backlit display increases visibility in low-light situations
  • Hypoglycemic Alert: Audio alert is designed to improve safety
  • qcProGuard™: Control solution test notifications provides additional quality control
  • ProGrip™: Rubberized-case design for secure handling
  • Strip release button - no need to touch used stripsUses regular AAA batteries
  • For in vitro Diagnostic Use Only
  • Critically ill patients should not be tested with the blood glucose meter
  • Not for use with neonates
Specifications
Sample Type Capillary Whole Blood
Type Blood Glucose Meter
Sample Size 0.5 μL