Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARKRAY USA Assure™ Platinum Test Strips

Catalog No. 22355168
Change view
Click to view available options
Quantity:
100 Pk.
50 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-355-168 100 Pk.
22-355-167 50 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-355-168 Supplier ARKRAY USA Supplier No. ARK 500100
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Assure™ Platinum Test Strips for use with the assure platinum meter only.

  • Sample Size: 0.5 μL
  • Chesmitry: Glucose Oxidase
  • Test Time: 7 Seconds
  • Open Bottle Shelf Life: 3 Months From Date Of Opening
  • For use with The Assure Platinum Meter Only
  • For in vitro Diagnostic Use Only
  • Critically ill patients should not be tested with the blood glucose meter
  • Not for use with neonates

Specifications

Color Code White
Clia Complexity Waived
Format Tube
Type Test Strips
Quantity 100 Pk.
Sample Size 0.5 μL
Test Time 7 sec.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.