Description
- Shingle pleat design prevents fluid pooling and cross-contamination
- Made from high-quality materials for reliable protection without compromising breathability
- Adjustable nosepiece ensures a comfortable, customized fit
Ultrasonically sealed, soft, stretchy, latex-free earloops prevent pulling and irritation - Available in all 3 levels of ASTM protection
Specifications
Specifications
| Product Type | Medical/Procedural Face Mask |
| ASTM Level | ASTM Level 1 |
| Color | Pink |
| Fastening Type | Earloops |
| Sterility | Non-sterile |
| Style | Pleated |
| Disposable | Single-use |
| Latex Free | Yes |