Learn More
Medicom™ AssureMask Balance™ Procedure Earloop Face Masks, ASTM Level 1
Catalog No. 19910760 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
Pink
White
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-910-760 Yellow
19-910-757 White
19-910-758 Blue
19-910-759 Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-910-760 Supplier Medicom™ Supplier No. 205417
Add to Cart
Add to Cart
This item is not returnable. View return policy
AssureMask Balance™ Procedure Earloop Face Masks, ASTM Level 1 provide healthcare professionals and other frontline workers with the ideal balance between the protection they need and the breathability and comfort they want. Made from high-quality materials.
  • Shingle pleat design prevents fluid pooling and cross-contamination
  • Made from high-quality materials for reliable protection without compromising breathability
  • Adjustable nosepiece ensures a comfortable, customized fit
    Ultrasonically sealed, soft, stretchy, latex-free earloops prevent pulling and irritation
  • Available in all 3 levels of ASTM protection
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 1
Color Yellow
Fastening Type Earloops
Sterility Non-sterile
Style Pleated
Disposable Single-use
Latex Free Yes
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov