Learn More
Medicom™ AssureMask™ Balance™ Earloop Face Masks
Catalog No. p-7237333 Shop All Medicom Products
Change view
Click to view available options
Color:
Blue
Pink
White
Yellow
ASTM Level:
ASTM Level 2
ASTM Level 3
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color ASTM Level
19-910-740 Blue ASTM Level 2
17-000-546 Pink ASTM Level 2
17-000-545 White ASTM Level 2
17-000-271 Yellow ASTM Level 2
17-000-544 Blue ASTM Level 3
17-000-548 Pink ASTM Level 3
17-000-547 White ASTM Level 3
17-000-549 Yellow ASTM Level 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-910-740 Supplier Medicom™ Supplier No. 205515
Add to Cart
Add to Cart
This item is not returnable. View return policy
Face masks provide healthcare professionals and other frontline workers with the ideal balance between protection and comfort without gapping or irritation.
  • Shingle pleat design prevents fluid pooling and cross-contamination
  • Adjustable nosepiece ensures a comfortable, customized fit
  • Ultrasonically sealed, soft, stretchy earloops are free of natural rubber latex, preventing pulling and irritation
  • Made from high-quality materials that provide reliable protection without compromising breathability
  • Assorted colors available
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 2
Color Blue
Fastening Type Earloops
Sterility Non-sterile
Style Pleated
Disposable Single-use
Latex Free Yes