Learn More
Medicom™ AssureMask Precision™ Care Face Mask with SureView™ Technology
Catalog No. 17000572 Shop All Medicom Products
Change view
Click to view available options
Color:
White
ASTM Level:
ASTM Level 1
Catalog No. Color ASTM Level
17-000-572 White ASTM Level 1
1 options
This item is not returnable. View return policy
Catalog No. 17-000-572 Supplier Medicom™ Supplier No. 206914
  • $164.31
    $142.65 / Case of 300
    Save $21.66
    13% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Tie-on face masks offer ASTM Level 1 protection while providing fog-free view with a personalized fit made especially for sensitive skin.
  • Made with SureView™ technology to ensure an unobstructed view of the operative field
  • Made from high quality materials that enhance breathability
  • Feature soft, ample-length ties, an adjustable nose piece, and a soft inner lining to ensure a comfortable, customized fit
  • Feature a breathable cellulose inner layer that remains cool on skin
  • Unique wicking technology keeps even the most sensitive faces dry and comfortable.
Specifications
Product Type General Purpose Face Mask
ASTM Level ASTM Level 1
Color White
Fastening Type Ties
Material Spunbond Polypropylene
Size Standard
Sterility Non-sterile
Style Pleated
Disposable Single-use
Filter Efficiency ≥95% (PFE), ≥95% (BFE)
Latex Free Yes
Model 206914
Packaging Quantity 50 Masks/Box, 6 Boxes/Cs.