Learn More
Medicom™ AssureMask Valiant™ Earloop Face Masks
Catalog No. 17000561 Shop All Medicom Products
This item is not returnable. View return policy
Catalog No. 17-000-561 Supplier Medicom™ Supplier No. 205917
Add to Cart
Add to Cart
This item is not returnable. View return policy
Procedure earloop face masks provide ASTM Level 1, 2 or 3 protection as well as optimal comfort and breathability.
  • Non-woven spunbond polypropylene outer layer provides maximum protection against fluid penetration
  • High-quality media ensure optimal filtration and breathability
  • Inner layer is made of dye-free non-woven spunbond polypropylene that will not lint or tear during use
  • Ample-length earloops are soft, strong, free of natural rubber latex, ultrasonically sealed, and attached to the outside of the mask to prevent pulling and irritation
  • Adjustable, enclosed, plastic-covered, iron nosepiece provides customized protection and comfort
  • Shingle pleat design prevents fluid pooling and cross-contamination
  • Made in North America to meet the highest quality and regulatory standards
  • Available in assorted colors
Specifications
Product Type Medical/Procedural Face Mask
Fastening Type Earloops
Material Spunbond Polypropylene
Size Standard
Sterility Non-sterile
Style Pleated
Disposable Single-use
Filter Efficiency ≥98% (PFE), ≥98% (BFE)
Latex Free Yes
Model 205917
Packaging Quantity 50 Masks/Box, 10 Boxes/Cs.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov