Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ AssureTouch™ Agile™ Nitrile Medical Examination Gloves with Textured Fingertips

Catalog No. 17101083 Shop All Medicom Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-101-083 X-Small
17-101-084 Small
17-101-085 Medium
17-101-086 Large
17-101-087 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-101-083 Supplier Medicom™ Supplier No. 9998A
Add to Cart
Add to Cart
This item is not returnable. View return policy

AssureTouch™ Agile™ Nitrile Medical Examination Gloves with Textured Fingertips. This nitrile glove offers optimal puncture and tear resistance without compromising dexterity or tactility. Textured fingertips ensure a secure grip, even when wet. Chemo tested.

  • These nitrile gloves offer superior puncture, tear and chemical resistance without compromising dexterity or tactility.
  • Textured fingertips ensure a secure grip, even when wet.
  • Chemo tested.

Specifications

Chlorinated No
Accelerator Free No
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Latex Free Yes
Powdered Powder Free
Size X-Small
Sterility Non-sterile
FDA 510 (k) Yes
Glove Cuff Style Beaded
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.