Learn More
Ritmed™ AssureWear™ Chemo Tested VersaGown™ with Flexneck™ Technology
Catalog No. 19910754 Shop All Medicom Products
Change view
Click to view available options
Size:
Large
X-Large
Color:
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19-910-754 Large Yellow
19-910-755 X-Large Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-910-754 Supplier Ritmed™ Supplier No. A69951
Add to Cart
Add to Cart
This item is not returnable. View return policy
Chemo tested VersaGown™ with Flexneck™ Technology provides AAMI Level 2 protection against cross-contamination for health professionals and patients.
  • High quality infection control
  • Quick and easy donning and removal while maintaining exceptional protection and comfort
  • Poly-coated and made with thumb loop cuffs to prevent sleeves from riding up and to ensure complete wrist coverage under exam gloves
  • Flexneck™ technology provides highly flexible gown closure
  • Design allows health care professionals to don protective apparel with a quick over-the-head motion eliminating the risk of untied gaping gowns that can slip and expose skin
Specifications
Size Large
Gender Unisex
No. of Pockets None
Product Type Chemotherapy Gown
Material Polyethylene, Polypropylene
Color Yellow
Closure Type Over-the-Head
Disposable Single-use
Sterility Non-sterile
Cuff Style Thumb Loop
Latex Free Yes
Model A69951
Sleeve Style Long Sleeve
Includes Left Side Belt
Certifications/Compliance AAMI Level 2, ASTM D6978, Medical Device Class 1
Autoclavable Not Autoclavable
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov