Learn More
Medicom™ AssureWear™ Chemotherapy Tested VersaGown™, Flexneck™ Technology
Catalog No. 19910855 Shop All Medicom Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Color:
Blue
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19-910-855 Small Blue
19-910-856 Medium Blue
19-910-857 Large Blue
19-910-858 X-Large Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-910-855 Supplier Medicom™ Supplier No. A69949
Add to Cart
Add to Cart
This item is not returnable. View return policy
AssureWear™ Chemo Tested VersaGown™ - Fast and easy donning to instantly provide complete coverage. Closure ensures safety by allowing gown to tear away from the body, preventing self-contamination. Poly coated and made with thumb loop cuffs.
  • VersaGown™ with Flexneck™ technology significantly reduces cross-contamination versus standard gowns.
  • Designed using clinician feedback and independent clinical testing.
  • Provides nearly two-fold reduction in contamination of hand and wrist areas
  • Helps ensure instant compliance—even without training.
  • Innovative design allows freedom of movement without compromising protection.
  • Flexneck™ creates the widest neck opening on the market and has ultrasonically sealed seams to prevent tearing during donning/doffing.
  • Winner of the Vizient Innovative Technology Award.
Specifications
Size Small
Product Type Chemotherapy Gown
Gender Unisex
No. of Pockets None
Color Blue
Disposable Single-use
Sterility Non-sterile
Cuff Style Thumb Loop
Latex Free Yes
Sleeve Style Long Sleeve
Certifications/Compliance AAMI Level 2
Autoclavable Not Autoclavable