Learn More
Ritmed™ AssureWear Over-the-head Isolation Gown, AAMI Level 2
Catalog No. 17000575 Shop All Medicom Products
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-000-575 Large
17-000-576 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-000-575 Supplier Ritmed™ Supplier No. A69977
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ultrasonically sealed, over-the-head isolation gowns offer AAMI PB70 Level 2 protection and are easy to don and doff.
  • Ultrasonically sealed over-the-head neck closure
  • Allows safe and easy donning and doffing
  • Offers effective barrier performance for low fluid level procedures with AAMI PB70 Level 2 rating
  • Full back closure provides complete coverage for optimal protection
  • Elastic, thumb loop cuffs prevent sleeves from riding up
  • Non-woven SMMS fabric keeps wearers cool and comfortable
  • Flat belt on left hand side ensures a personalized fit
Specifications
Product Type Isolation Gown
No. of Pockets None
Size Large
Gender Unisex
Material SMMS Polypropylene
Color Yellow
Disposable Single-use
Sterility Non-sterile
Cuff Style Thumb Loop
Latex Free Yes
Sleeve Style Long Sleeve
Certifications/Compliance AAMI PB70 Level 2
Autoclavable Not Autoclavable