Learn More
Ritmed™ AssureWear™ SMMS VersaGown™ Isolation Gowns with Flexneck™ Technology, Thumb Loops
Catalog No. 19910749 Shop All Medicom Products
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-910-749 Large
19-910-750 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-910-749 Supplier Ritmed™ Supplier No. A69911T
Add to Cart
Add to Cart
This item is not returnable. View return policy
Disposable SMMS polypropylene isolation gowns with thumb loops are designed to protect health professionals and patients from cross infection. Patented Flexneck™ technology for fast and easy donning and removal.
  • Made of SMMS non-woven material with full back closure
  • Features flat belt elastic cuffs with thumb loops and tie-on neck closure
Specifications
Size Large
Gender Unisex
No. of Pockets None
Product Type Isolation Gown
Material SMMS Polypropylene
Color Yellow
Closure Type Neck Ties
Disposable Single-use
Sterility Non-sterile
Cuff Style Thumb Loop
Latex Free Yes
Model A69911-T
Sleeve Style Long Sleeve
Includes Flat Front Belt
Certifications/Compliance AAMI PB70 level 1, Medical Device Class 1
Autoclavable Not Autoclavable
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov