Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ AssureWear™ Blue SMS Lab Coats

Catalog No. 19910810 Shop All Medicom Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3X-Large
4X-Large
7X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-910-810 4X-Large
19-910-805 Medium
19-910-806 Large
19-910-807 X-Large
19-910-809 3X-Large
19-910-811 7X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-910-810 Supplier Medicom™ Supplier No. A8038
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Non-woven SMS lab coat with knitted cuffs is designed to protect healthcare professionals, lab technicians, and other wearers from fluids and cross-contamination.

  • Non-woven 3-layer SMS
  • Knit cuffs
  • Knit collar

Specifications

Product Type General Purpose Lab Coat
Size 4X-Large
Gender Unisex
Material SMS
Color Blue
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Style Knee-length
No. of Pockets None
Collar Type Knit
Latex Free Yes
Model A8038
For Use With (Application) Healthcare, Laboratories
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.