Learn More
Ritmed™ AssureWear™ VersaGown™ with Flexneck™ Technology, AAMI Level 2
Catalog No. 19910753 Shop All Medicom Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-910-753 2X-Large
19-910-751 Large
19-910-752 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-910-753 Supplier Ritmed™ Supplier No. A69963
Add to Cart
Add to Cart
This item is not returnable. View return policy
Isolation gowns offer AAMI Level 2 protection to reduce risk of cross-contamination among health professionals and patients. Patented Flexneck™ technology provides fast and easy donning and removal.
  • Designed using clinician feedback and independent clinical testing
  • Provides nearly two-fold reduction in contamination of hand and wrist areas
  • Helps ensure instant compliance, even without training
  • Innovative design allows freedom of movement without compromising protection
  • Patented Flexneck™ technology allows fast and easy donning and removal while providing exceptional protection and comfort
  • Innovative Flexneck™ closure quickly and easily passes over the head to instantly provide complete coverage and eliminate the risk posed by untied gaping gowns that can slip and expose skin
  • Flexneck™ closure also allows easy doffing that allows gown to swiftly yet gently tear away from the body preventing self-contamination
Specifications
Size 2X-Large
Gender Unisex
No. of Pockets None
Product Type Isolation Gown
Material SMMS Polypropylene
Color Yellow
Closure Type Over-the-Head
Disposable Single-use
Sterility Non-sterile
Cuff Style Thumb Loop
Latex Free Yes
Model A69963
Sleeve Style Long Sleeve
Includes Left Side Belt
Certifications/Compliance AAMI Level 2, Medical Device Class 1
Autoclavable Not Autoclavable
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov