Learn More
Ritmed™ AssureWear VersaGown™ Lightweight Isolation Gown with Flexneck Technology, AAMI Level 2
Catalog No. 17000573 Shop All Medicom Products
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-000-573 Large
17-000-574 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-000-573 Supplier Ritmed™ Supplier No. A69921
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lightweight isolation gown features Flexneck™ closure for easy and quick doffing to prevent self-contamination.
  • Significantly reduces cross-contamination versus standard gowns
  • Designed using clinician feedback and independent clinical testing
  • Provides nearly two-fold reduction in contamination of hand and wrist areas
  • Helps ensure instant compliance, even without training
  • Innovative design allows freedom of movement without compromising protection
  • Flexneck™ creates the widest neck opening on the market and has ultrasonically sealed seams to prevent tearing during donning and doffing
  • 100 gowns per case
Specifications
Product Type Isolation Gown
Size Large
Gender Unisex
No. of Pockets None
Material SMMS Polypropylene
Color Yellow
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Latex Free Yes
Sleeve Style Long Sleeve
Certifications/Compliance AAMI Level 2
Autoclavable Not Autoclavable