Learn More
Waters Corp Waters Ball and Seat Outlet Check Valve, 225 μL
Catalog No. 50819013 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This prefabricated valve unit saves installation time and reduces the handling of small parts, as compared with the valve replacement process using the corresponding valve rebuild kit. The valve is convenient and easy to install in Waters 515 and 600 pumps, as well as the LC Module 1 system. All Waters check valves are leak-tested at the factory and assembled in a clean environment.
  • Saves installation time and reduces the handling of small parts
  • Convenient and easy to install in Waters 515 and 600 pumps, as well as the LC Module 1 system
  • Leak-tested at the factory
  • Assembled in a clean environment
  • Units per package: 1
  • Annual replacement schedule
Catalog No. 50-819-013 Supplier Waters Corp Supplier No. WAT025216
This prefabricated valve unit saves installation time and reduces the handling of small parts, as compared with the valve replacement process using the corresponding valve rebuild kit. The valve is convenient and easy to install in Waters 515 and 600 pumps, as well as the LC Module 1 system. All Waters check valves are leak-tested at the factory and assembled in a clean environment.
  • Saves installation time and reduces the handling of small parts
  • Convenient and easy to install in Waters 515 and 600 pumps, as well as the LC Module 1 system
  • Leak-tested at the factory
  • Assembled in a clean environment
  • Units per package: 1
  • Annual replacement schedule