Learn More
Waters Corp Check Valve Cartridge Housing, Stainless Steel, Outlet
Catalog No. 50792595 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is a replacement stainless steel check valve cartridge housing designed for the outlet side of various HPLC systems. It is compatible with 2535, 2545, 2555 pumps, and the ACQUITY H-Class Quaternary Solvent Manager.
  • Made from stainless steel.
  • Designed for the outlet.
  • Replacement part for high performance liquid chromatography systems.
  • Compatible with 2535, 2545, and 2555 pumps.
  • Compatible with ACQUITY H-Class Quaternary Solvent Manager.
  • Does not include a replacement check valve cartridge.
  • Intended for replacement as required.
Catalog No. 50-792-595 Supplier Waters Corp Supplier No. 700001530
This is a replacement stainless steel check valve cartridge housing designed for the outlet side of various HPLC systems. It is compatible with 2535, 2545, 2555 pumps, and the ACQUITY H-Class Quaternary Solvent Manager.
  • Made from stainless steel.
  • Designed for the outlet.
  • Replacement part for high performance liquid chromatography systems.
  • Compatible with 2535, 2545, and 2555 pumps.
  • Compatible with ACQUITY H-Class Quaternary Solvent Manager.
  • Does not include a replacement check valve cartridge.
  • Intended for replacement as required.