Learn More
Waters Corp Waters Vent Valve Cartridge Assembly, Tubing, UPLC System
Catalog No. 50201705 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This vent valve cartridge assembly serves as a replacement component for various UPLC systems and solvent managers.
  • Replacement vent valve cartridge assembly
  • Designed for UPLC system applications
  • Compatible with ACQUITY I-Class Binary Solvent Managers
  • Compatible with ACQUITY Isocratic Solvent Manager
  • Compatible with PATROL Sample Manager
  • Compatible with ACQUITY UPLC I-Class System
  • Compatible with ACQUITY UPLC ISM
  • Compatible with ACQUITY UPLC BSM
  • Compatible with ACQUITY UPLC Process Sample Manager
  • Supplied as 1 unit per package
  • Features tubing type construction
  • Replaced as required for optimal performance
Catalog No. 50-201-705 Supplier Waters Corp Supplier No. 700006052
This vent valve cartridge assembly serves as a replacement component for various UPLC systems and solvent managers.
  • Replacement vent valve cartridge assembly
  • Designed for UPLC system applications
  • Compatible with ACQUITY I-Class Binary Solvent Managers
  • Compatible with ACQUITY Isocratic Solvent Manager
  • Compatible with PATROL Sample Manager
  • Compatible with ACQUITY UPLC I-Class System
  • Compatible with ACQUITY UPLC ISM
  • Compatible with ACQUITY UPLC BSM
  • Compatible with ACQUITY UPLC Process Sample Manager
  • Supplied as 1 unit per package
  • Features tubing type construction
  • Replaced as required for optimal performance