Learn More
Waters Corp Waters Corporation Diffuser Assembly, Solvent Filter, HPLC
Catalog No. 50818985 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This diffuser assembly functions as a solvent filter for High-Performance Liquid Chromatography (HPLC) systems and can also be used as a helium diffuser. It is designed to be a replacement filter for the seal wash inlet line on Alliance systems and can also be found in a solvent bottle cap kit.
  • Functions as a replacement filter for seal wash inlet lines.
  • Can be used as a filter within solvent bottle cap kits.
  • Disperses helium gas into the mobile phase when used as a helium diffuser assembly.
  • Suitable for High-Performance Liquid Chromatography (HPLC) systems.
Catalog No. 50-818-985 Supplier Waters Corp Supplier No. WAT007272
This diffuser assembly functions as a solvent filter for High-Performance Liquid Chromatography (HPLC) systems and can also be used as a helium diffuser. It is designed to be a replacement filter for the seal wash inlet line on Alliance systems and can also be found in a solvent bottle cap kit.
  • Functions as a replacement filter for seal wash inlet lines.
  • Can be used as a filter within solvent bottle cap kits.
  • Disperses helium gas into the mobile phase when used as a helium diffuser assembly.
  • Suitable for High-Performance Liquid Chromatography (HPLC) systems.