Learn More
Waters Corp Extension Loop, 100 μL, Bio, HPS
Catalog No. 502392624 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 100 μL biocompatible extension loop utilizes MaxPeak HPS Technology. It is designed for use with ACQUITY Premier and Arc Premier Systems, allowing for additional sample volume to be drawn and held for injection.
  • 100 μL biocompatible extension loop
  • Utilizes MaxPeak HPS Technology
  • Used with ACQUITY Premier and Arc Premier Systems
  • Allows additional volume of sample to be drawn and held for injection
Catalog No. 50-239-2624 Supplier Waters Corp Supplier No. 700012826
This 100 μL biocompatible extension loop utilizes MaxPeak HPS Technology. It is designed for use with ACQUITY Premier and Arc Premier Systems, allowing for additional sample volume to be drawn and held for injection.
  • 100 μL biocompatible extension loop
  • Utilizes MaxPeak HPS Technology
  • Used with ACQUITY Premier and Arc Premier Systems
  • Allows additional volume of sample to be drawn and held for injection