Learn More
Cole-Parmer Labglass BP-1755-000 Filtration Assembly, 47mm dia, 300 mL
Supplier:  Cole-Parmer 3450900
Encompass_Preferred
Labglass™ filtration assemblies are designed for HPLC, particulate, and microbiological contamination filtrations. Sintered glass support provides even, flat support structure for membrane filters. Available in 47-mm or 90-mm filter sizes. Order Labglass™ filtering flasks separately.
Catalog No. NC1485453