Learn More
Waters Corp Waters Frit Assembly, 0.2 μm, 2.1 mm Column, 3-Pack
Catalog No. 50179750 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This frit assembly serves as a replacement for the inlet/outlet frits in 2.1 mm I.D. ACQUITY UPLC Columns. It is designed for straightforward assembly and aims to uphold the original quality, efficiency, and peak performance of your instrument. These components are rigorously tested and certified for optimal integration with systems, ensuring consistent operation and reliable results.
  • Replaces inlet/outlet frits for 2.1 mm I.D. ACQUITY UPLC columns.
  • Designed for easy assembly with the original column.
  • Aims to deliver the same quality, efficiency, and peak performance as the original part.
  • Ensures consistent instrument operation, accurate and reproducible results, and continued system compliance.
  • Compatible with CM, CH, and CHC column managers.
Catalog No. 50-179-750 Supplier Waters Corp Supplier No. 700003776
This frit assembly serves as a replacement for the inlet/outlet frits in 2.1 mm I.D. ACQUITY UPLC Columns. It is designed for straightforward assembly and aims to uphold the original quality, efficiency, and peak performance of your instrument. These components are rigorously tested and certified for optimal integration with systems, ensuring consistent operation and reliable results.
  • Replaces inlet/outlet frits for 2.1 mm I.D. ACQUITY UPLC columns.
  • Designed for easy assembly with the original column.
  • Aims to deliver the same quality, efficiency, and peak performance as the original part.
  • Ensures consistent instrument operation, accurate and reproducible results, and continued system compliance.
  • Compatible with CM, CH, and CHC column managers.