Learn More
Waters Corp Waters Tubing Assembly, Detector Inlet, PEEK, 0.004 in. ID, 8.5 in. L
Catalog No. NC1672451 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is a replacement tubing assembly designed for detector inlets. It is constructed from PEEK material with a 0.004 in. internal diameter and an 8.5 in. length, and includes a PEEK nut. This assembly is suitable for UPLC systems and is specifically used to connect to ACQUITY TUV Flow Cells and to attach the column outlet to the detector during column installation.
  • Replacement tubing assembly for detector inlets.
  • Constructed from PEEK material.
  • Features a 0.004 in. internal diameter.
  • Measures 8.5 in. in length.
  • Includes a PEEK nut.
  • Connects to ACQUITY TUV Flow Cells.
  • Attaches the column outlet to the detector during column installation.
  • Suitable for UPLC systems.
Catalog No. NC1672451 Supplier Waters Corp Supplier No. 700009972
This is a replacement tubing assembly designed for detector inlets. It is constructed from PEEK material with a 0.004 in. internal diameter and an 8.5 in. length, and includes a PEEK nut. This assembly is suitable for UPLC systems and is specifically used to connect to ACQUITY TUV Flow Cells and to attach the column outlet to the detector during column installation.
  • Replacement tubing assembly for detector inlets.
  • Constructed from PEEK material.
  • Features a 0.004 in. internal diameter.
  • Measures 8.5 in. in length.
  • Includes a PEEK nut.
  • Connects to ACQUITY TUV Flow Cells.
  • Attaches the column outlet to the detector during column installation.
  • Suitable for UPLC systems.