Learn More
Waters Corp Waters Inlet Tubing Assembly, BioACQUITY, MP35N, 0.004 in ID
Catalog No. NC1487270 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This inlet tubing assembly is a replacement part designed for BioACQUITY systems that utilize a 30 cm Column Heater/Cooler.
  • Manufactured from MP35N material.
  • Features an internal diameter of 0.004 in.
  • Designed for UPLC system applications.
Catalog No. NC1487270 Supplier Waters Corp Supplier No. 430002575
This inlet tubing assembly is a replacement part designed for BioACQUITY systems that utilize a 30 cm Column Heater/Cooler.
  • Manufactured from MP35N material.
  • Features an internal diameter of 0.004 in.
  • Designed for UPLC system applications.