Learn More
Hach Company SL1000 - PPA Portable Parallel Analyzer- Portable Colorimeter with USB
Catalog No. 501997344 Shop All Hach Company Products
Encompass
Complete more tests on site, get the results you need faster, allowing you to visit more sites in each shift. Perform up to four colorimetric and two probe-based measurements in parallel 75% faster than with other methods.The Hach SL1000 Portable Parallel Analyzer (PPA) performs the same tests with less than half the manual steps. Get highly accurate results, with less opportunity for errors, in a fraction of the time. Up to six parameters, tested in parallel. The SL1000 is a portable colorimeter that provides 75% faster testing in chloramination applications by using Chemkey reagents instead of powder pillows. All chemicals and processes are entirely contained inside the Chemkey.
Catalog No. 50-199-7344 Supplier Hach Company Supplier No. 9430000
Add to Cart
Add to Cart
Request a Quote
Complete more tests on site, get the results you need faster, allowing you to visit more sites in each shift. Perform up to four colorimetric and two probe-based measurements in parallel 75% faster than with other methods.The Hach SL1000 Portable Parallel Analyzer (PPA) performs the same tests with less than half the manual steps. Get highly accurate results, with less opportunity for errors, in a fraction of the time. Up to six parameters, tested in parallel. The SL1000 is a portable colorimeter that provides 75% faster testing in chloramination applications by using Chemkey reagents instead of powder pillows. All chemicals and processes are entirely contained inside the Chemkey.