Learn More
Waters Corp Mixer, 100 μL, UPLC
Supplier: Waters Corp 700005762
This optional 100μL mixer assembly is designed to enhance the mixing of large volume injections, particularly those with high organic content, before chromatographic separation. It is compatible with UPLC systems and aids in the transfer of legacy HPLC methods.
- Enhances mixing of large volume injections with high organic content.
- Supports improved mixing prior to separation.
- Aids in the transfer of legacy HPLC methods.
- Compatible with H-Class Quaternary Solvent Manager.
- Compatible with ACQUITY UPLC H-Class System’s Sample Manager – FTN.