Learn More
Waters Corp ACQUITY UPLC Mixer, 18K, 100 μL
Catalog No. 50201653 Shop All Waters Corporation Products
Encompass_Preferred
This optional mixer has a 100 μL capacity and is designed for use with ACQUITY H-Class/I-Class Binary Solvent Manager systems.
  • 1 package
  • Mixer type
  • UPLC system type
  • Compatible with H-Class and I-Class systems
  • Replacement as required
  • Component of Binary Solvent Manager (BSM)
Catalog No. 50-201-653 Supplier Waters Corp Supplier No. 205000854
This optional mixer has a 100 μL capacity and is designed for use with ACQUITY H-Class/I-Class Binary Solvent Manager systems.
  • 1 package
  • Mixer type
  • UPLC system type
  • Compatible with H-Class and I-Class systems
  • Replacement as required
  • Component of Binary Solvent Manager (BSM)