Learn More
Waters Corp Waters Corporation Mixer Assembly, 2545Q, Quaternary Gradient Module Component
Catalog No. 50792995 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This mixer assembly is designed for use with the 2545 Quaternary Gradient Module. It plays a crucial role in mixing solvents to enhance the compositional ripple in chromatographic systems, ensuring consistent and stable solvent compositions for analytical processes.
  • Integrates with the 2545 Quaternary Gradient Module.
  • Enhances compositional ripple of solvents.
  • Contributes to consistent and stable solvent composition.
  • Essential component for chromatographic systems.
  • Categorized as a spare part for maintenance.
Catalog No. 50-792-995 Supplier Waters Corp Supplier No. 700004436
This mixer assembly is designed for use with the 2545 Quaternary Gradient Module. It plays a crucial role in mixing solvents to enhance the compositional ripple in chromatographic systems, ensuring consistent and stable solvent compositions for analytical processes.
  • Integrates with the 2545 Quaternary Gradient Module.
  • Enhances compositional ripple of solvents.
  • Contributes to consistent and stable solvent composition.
  • Essential component for chromatographic systems.
  • Categorized as a spare part for maintenance.