Learn More
Waters Corp Waters Mixer, 50 μL, Zirconia, UPLC
Catalog No. NC9623521 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This mixer is designed for use with ACQUITY UPLC systems, featuring a 50 μL capacity and durable zirconia construction. It functions as a standard mixer and is suitable for high-pressure applications.
  • 50 μL capacity for precise mixing.
  • Constructed with zirconia for durability and chemical resistance.
  • Standard mixer type for general laboratory use.
  • Optimized for ACQUITY UPLC systems.
  • Functions as a component within the binary solvent manager.
  • Suitable for high pressure applications as a filter mixer.
  • Supplied as one unit per package.
Catalog No. NC9623521 Supplier Waters Corp Supplier No. 700002911
This mixer is designed for use with ACQUITY UPLC systems, featuring a 50 μL capacity and durable zirconia construction. It functions as a standard mixer and is suitable for high-pressure applications.
  • 50 μL capacity for precise mixing.
  • Constructed with zirconia for durability and chemical resistance.
  • Standard mixer type for general laboratory use.
  • Optimized for ACQUITY UPLC systems.
  • Functions as a component within the binary solvent manager.
  • Suitable for high pressure applications as a filter mixer.
  • Supplied as one unit per package.