Learn More
Hach Company Dissolved Iron Chemkey Reagents (box of 25)
Supplier:  Hach Company 8636000
Encompass
Dissolved Iron Chemkeys, range 0.05 - 3.00 mg/L Fe. For SL1000 Portable Parallel Analyzer (PPA). Box of 25.
Catalog No. 50-199-7271